missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromopyrimidine, 97%, ACROS Organics™

Brand:  Acros Organics 368580050

UNSPSC : 12350000

Code : 29

Additional Details : CAS Number : 7752-82-1 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10512252

  • £66.30 / 5g


Estimated Shipment
17-10-2017
Add to basket


Specifications

Chemical Identifiers

2-Amino-5-bromopyrimidine
7752-82-1
100.0
97%
174
97%
96.0
≥96.0%
C4H4BrN3
MFCD00012341

Specifications

2-Amino-5-bromopyrimidine
7752-82-1
100.0
Glass bottle
≥96.0%
230° to 244°C
C4H4BrN3
5g
25, IV, 2125
97%
96.0
Authentic
Crystalline Powder, Crystals or Flakes
97%
White to light beige
174
MFCD00012341
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Amino-5-bromopyrimidine

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Documentation

Technical Documents