missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromo-4-(trifluoromethyl)pyrimidine, 95%, Thermo Scientific™

Catalog Number 15438946
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
935534-47-7
C5H3BrF3N3
241.999
MFCD09261254
FAMGPURZLOTOKD-UHFFFAOYSA-N
5-bromo-4-trifluoromethyl pyrimidin-2-amine, 5-bromo-4-trifluoromethyl-pyrimidin-2-ylamine, 5-bromo-4-trifluoromethyl-2-pyrimidinamine, ksc672m4h, 2-amino-5-bromo-4-trifluoromethylpyrimidine, 5-bromo-4-trifluoromethyl-2-pyrimidineamin, 5-bromo-4-trifluoromethyl-2-pyrimidylamine, 2-amino-5-bromo-4-trifluoromethyl pyrimidine, 2-pyrimidinamine, 5-bromo-4-trifluoromethyl, 5-bromo-4-trifluoromethyl pyrimidine-2-ylamine
45382152
5-bromo-4-(trifluoromethyl)pyrimidin-2-amine
C1=C(C(=NC(=N1)N)C(F)(F)F)Br
This item is not returnable. View return policy
935534-47-7
C5H3BrF3N3
241.999
MFCD09261254
FAMGPURZLOTOKD-UHFFFAOYSA-N
5-bromo-4-trifluoromethyl pyrimidin-2-amine, 5-bromo-4-trifluoromethyl-pyrimidin-2-ylamine, 5-bromo-4-trifluoromethyl-2-pyrimidinamine, ksc672m4h, 2-amino-5-bromo-4-trifluoromethylpyrimidine, 5-bromo-4-trifluoromethyl-2-pyrimidineamin, 5-bromo-4-trifluoromethyl-2-pyrimidylamine, 2-amino-5-bromo-4-trifluoromethyl pyrimidine, 2-pyrimidinamine, 5-bromo-4-trifluoromethyl, 5-bromo-4-trifluoromethyl pyrimidine-2-ylamine
45382152
5-bromo-4-(trifluoromethyl)pyrimidin-2-amine
C1=C(C(=NC(=N1)N)C(F)(F)F)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 242
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Amino-5-bromo-4-(trifluoromethyl)pyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'