missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromo-4-hydroxy-6-methylpyrimidine, 99%, Thermo Scientific™

Catalog Number p-7025368
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
6307-35-3
C5H6BrN3O
204.027
MFCD00006092
ADLWOFHKMXUDKF-UHFFFAOYSA-N
2-amino-5-bromo-6-methyl-4-pyrimidinol, 2-amino-5-bromo-4-hydroxy-6-methylpyrimidine, 2-amino-5-bromo-6-methylpyrimidin-4-ol, 2-amino-5-bromo-6-methyl-4 3h-pyrimidinone, abmp, pubchem6984, acmc-20ao4q, 2-amino-5-bromo-6-methylpyrimidin-4 1h-one, 5-bromo-6-methylisocytosine, chembl80800
80567
2-amino-5-bromo-6-methyl-1H-pyrimidin-4-one
CC1=C(C(=O)N=C(N1)N)Br
This item is not returnable. View return policy
6307-35-3
C5H6BrN3O
204.027
MFCD00006092
ADLWOFHKMXUDKF-UHFFFAOYSA-N
2-amino-5-bromo-6-methyl-4-pyrimidinol, 2-amino-5-bromo-4-hydroxy-6-methylpyrimidine, 2-amino-5-bromo-6-methylpyrimidin-4-ol, 2-amino-5-bromo-6-methyl-4 3h-pyrimidinone, abmp, pubchem6984, acmc-20ao4q, 2-amino-5-bromo-6-methylpyrimidin-4 1h-one, 5-bromo-6-methylisocytosine, chembl80800
80567
2-amino-5-bromo-6-methyl-1H-pyrimidin-4-one
CC1=C(C(=O)N=C(N1)N)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point ∼240°C (decomposition)
Quantity 5 g
Beilstein 4959
Formula Weight 204.03
Percent Purity 99%
Chemical Name or Material 2-Amino-5-bromo-4-hydroxy-6-methylpyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'