missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromo-3-hydroxypyridine, 95%, Thermo Scientific Chemicals

Catalog Number 15428288
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
39903-01-0
C5H5BrN2O
189.01
MFCD09744143
YQADLKDQAXAIKW-UHFFFAOYSA-N
2-amino-5-bromo-3-pyridinol, 2-amino-5-bromo-3-hydroxypyridine, 2-amino-3-hydroxy-5-bromopyridine, 3-pyridinol, 2-amino-5-bromo, 2-amino-5-bromo-pyridin-3-ol, 2-amino-5-bromopyridine-3-ol, acmc-209j8w, 2-amino-5-bromopryidin-3-ol, 2-amino-3-hydroxy-5-bromo pyridine, 5-bromo-3-hydroxy-2-amino-pyridine
11694041
2-amino-5-bromopyridin-3-ol
NC1=NC=C(Br)C=C1O
This item is not returnable. View return policy
39903-01-0
C5H5BrN2O
189.01
MFCD09744143
YQADLKDQAXAIKW-UHFFFAOYSA-N
2-amino-5-bromo-3-pyridinol, 2-amino-5-bromo-3-hydroxypyridine, 2-amino-3-hydroxy-5-bromopyridine, 3-pyridinol, 2-amino-5-bromo, 2-amino-5-bromo-pyridin-3-ol, 2-amino-5-bromopyridine-3-ol, acmc-209j8w, 2-amino-5-bromopryidin-3-ol, 2-amino-3-hydroxy-5-bromo pyridine, 5-bromo-3-hydroxy-2-amino-pyridine
11694041
2-amino-5-bromopyridin-3-ol
NC1=NC=C(Br)C=C1O

2-Amino-5-bromo-3-hydroxypyridine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-bromo-3-hydroxypyridine is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 189.01
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Amino-5-bromo-3-hydroxypyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'