missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromo-2'-fluorobenzophenone, 95%, Thermo Scientific Chemicals

Catalog Number 15408478
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1479-58-9
C13H9BrFNO
294.12
MFCD00038380
XCOKDXNGCQXFCV-UHFFFAOYSA-N
2-amino-5-bromo-2'-fluorobenzophenone, 2-amino-2'-fluoro-5-bromobenzophenone, 2-amino-5-bromophenyl 2-fluorophenyl methanone, methanone, 2-amino-5-bromophenyl 2-fluorophenyl, 4-bromo-2-2-fluorobenzoyl aniline, 2-amino-5-bromophenyl 2-fluorophenyl ketone, 2-amino-5-bromo-2-fluorobenzophenone, 2-amino-5-bromo-phenyl-2-fluoro-phenyl-methanone, 2-amino-2-fluoro-5-bromobenzophenone, 2-amino-5-bromo-2'-fluoro benzophenone
73865
(2-amino-5-bromophenyl)-(2-fluorophenyl)methanone
NC1=CC=C(Br)C=C1C(=O)C1=CC=CC=C1F
This item is not returnable. View return policy
1479-58-9
C13H9BrFNO
294.12
MFCD00038380
XCOKDXNGCQXFCV-UHFFFAOYSA-N
2-amino-5-bromo-2'-fluorobenzophenone, 2-amino-2'-fluoro-5-bromobenzophenone, 2-amino-5-bromophenyl 2-fluorophenyl methanone, methanone, 2-amino-5-bromophenyl 2-fluorophenyl, 4-bromo-2-2-fluorobenzoyl aniline, 2-amino-5-bromophenyl 2-fluorophenyl ketone, 2-amino-5-bromo-2-fluorobenzophenone, 2-amino-5-bromo-phenyl-2-fluoro-phenyl-methanone, 2-amino-2-fluoro-5-bromobenzophenone, 2-amino-5-bromo-2'-fluoro benzophenone
73865
(2-amino-5-bromophenyl)-(2-fluorophenyl)methanone
NC1=CC=C(Br)C=C1C(=O)C1=CC=CC=C1F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 100°C to 104°C
Quantity 1 g
Formula Weight 294.12
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Amino-5-bromo-2'-fluorobenzophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'