missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(trifluoromethyl)pyridine, 97%, ACROS Organics™

Brand:  Acros Organics 442641000

Code : 29

Additional Details : CAS Number : 106447-97-6 Weight : 0.05010kg Transport : UN number : 2811 Chem class : 1 Pack group : III


Quantity 100mg

 View more versions of this product

Product Code. 11926441

  • £33.00 / 100mg


Estimated Shipment
13-02-2018
Add to basket


Specifications

Chemical Identifiers

2-Amino-4-(trifluoromethyl)pyridine
96.0
≥96.0%
C6H5F3N2
106447-97-6
100.0
97%
162.11

Specifications

2-Amino-4-(trifluoromethyl)pyridine
96.0
Authentic
≥96.0%
C6H5F3N2
162.11
106447-97-6
100.0
Powder, Solid or Crystalline
97%
White to pale brown
100mg
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Amino-4-(trifluoromethyl)pyridine

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS