missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-thiazoleacetic acid hydrochloride, 98%, Thermo Scientific™

Catalog Number p-7030456
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
66659-20-9
C5H7ClN2O2S
194.633
MFCD06658943
ZEWJJZKVQOMYKJ-UHFFFAOYSA-N
2-2-aminothiazol-4-yl acetic acid hydrochloride, 2-amino-4-thiazoleacetic acid hydrochloride, 2-2-aminothiazol-4-yl acetic acid hcl, 2-2-amino-4-thiazolyl acetic acid hydrochloride, 4-thiazoleacetic acid, 2-amino-, monohydrochloride, 2-2-aminothiazol-4-yl acetic acid,hydrochloride, 2-amino-1,3-thiazol-4-yl acetic acid hydrochloride, 2-2-amino-4-thiazolyl acetic acid hcl, 2-aminothiazol-4-ylacetic acid hydrochloride
13017673
2-(2-amino-1,3-thiazol-4-yl)acetic acid;hydrochloride
C1=C(N=C(S1)N)CC(=O)O.Cl
This item is not returnable. View return policy
66659-20-9
C5H7ClN2O2S
194.633
MFCD06658943
ZEWJJZKVQOMYKJ-UHFFFAOYSA-N
2-2-aminothiazol-4-yl acetic acid hydrochloride, 2-amino-4-thiazoleacetic acid hydrochloride, 2-2-aminothiazol-4-yl acetic acid hcl, 2-2-amino-4-thiazolyl acetic acid hydrochloride, 4-thiazoleacetic acid, 2-amino-, monohydrochloride, 2-2-aminothiazol-4-yl acetic acid,hydrochloride, 2-amino-1,3-thiazol-4-yl acetic acid hydrochloride, 2-2-amino-4-thiazolyl acetic acid hcl, 2-aminothiazol-4-ylacetic acid hydrochloride
13017673
2-(2-amino-1,3-thiazol-4-yl)acetic acid;hydrochloride
C1=C(N=C(S1)N)CC(=O)O.Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point ∽152°C (decomposition)
Quantity 5 g
Formula Weight 194.64
Percent Purity 98%
Chemical Name or Material 2-Amino-4-thiazoleacetic acid hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'