missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-tert-butylthiazole, 98%, Thermo Scientific Chemicals

Catalog Number p-7021683
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
74370-93-7
C7H12N2S
156.247
MFCD00051644
CUWZBHVYLVGOAB-UHFFFAOYSA-N
2-amino-4-tert-butylthiazole, 4-tert-butylthiazol-2-ylamine, 4-tert-butyl thiazol-2-amine, 4-tert-butyl-thiazol-2-ylamine, 2-amino-4-t-butylthiazole, 4-tert-butyl-1,3-thiazol-2-amine, 4-tert-butylthiazol-2-amine, 4-tert-butyl-1,3-thiazol-2-ylamine, 2-thiazolamine, 4-1,1-dimethylethyl, 4-tert-butyl-1,3-thiazole-2-ylamine
2734202
4-tert-butyl-1,3-thiazol-2-amine
CC(C)(C)C1=CSC(=N1)N
This item is not returnable. View return policy
74370-93-7
C7H12N2S
156.247
MFCD00051644
CUWZBHVYLVGOAB-UHFFFAOYSA-N
2-amino-4-tert-butylthiazole, 4-tert-butylthiazol-2-ylamine, 4-tert-butyl thiazol-2-amine, 4-tert-butyl-thiazol-2-ylamine, 2-amino-4-t-butylthiazole, 4-tert-butyl-1,3-thiazol-2-amine, 4-tert-butylthiazol-2-amine, 4-tert-butyl-1,3-thiazol-2-ylamine, 2-thiazolamine, 4-1,1-dimethylethyl, 4-tert-butyl-1,3-thiazole-2-ylamine
2734202
4-tert-butyl-1,3-thiazol-2-amine
CC(C)(C)C1=CSC(=N1)N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 98°C to 103°C
Quantity 1 g
Beilstein 114270
Formula Weight 156.26
Percent Purity 98%
Chemical Name or Material 2-Amino-4-tert-butylthiazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'