missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methylpyrimidine-5-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Catalog Number p-7036127
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
944401-55-2
C11H18BN3O2
235.09
MFCD18072554
SYJMHOBGFXCKRG-UHFFFAOYSA-N
52987906
4-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine
CC1=NC(N)=NC=C1B1OC(C)(C)C(C)(C)O1
This item is not returnable. View return policy
944401-55-2
C11H18BN3O2
235.09
MFCD18072554
SYJMHOBGFXCKRG-UHFFFAOYSA-N
52987906
4-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine
CC1=NC(N)=NC=C1B1OC(C)(C)C(C)(C)O1

2-Amino-4-methylpyrimidine-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-4-methylpyrimidine-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Synonym 4-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, 4-methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, 4-methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyrimidin-2-ylamine, 2-amino-4-methylpyrimidine-5-boronic acid pinacol ester, 4-methyl-2-aminopyrimidine-5-boronic acid pinacol ester, 2-amino-4-methylpyrimidin-5-yl boronic acid pinacol ester, 4-methyl-5-4, 4,5,5-tetramethyl 1,3,2-dioxaborolan-2-yl pyrimidine-2-ylamine, 4-methyl-5-4,4,5,5-tetramethyl 1,3,2-dioxaborolan-2-yl pyrimidine-2-ylamine
Solubility Information Slightly soluble in water.
Formula Weight 235.09
Percent Purity 96%
Chemical Name or Material 2-Amino-4-methylpyrimidine-5-boronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'