missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methylbenzonitrile, 97%, Thermo Scientific™

Catalog Number 10672273
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
26830-96-6
C8H8N2
132.17
MFCD00173706
LGNVAEIITHYWCG-UHFFFAOYSA-N
2-cyano-5-methylaniline, 3-amino-4-cyanotoluene, 2-amino-4-methyl-benzonitrile, 4-methyl-2-aminobenzonitrile, benzonitrile, 2-amino-4-methyl, 2-amino-4-methylbenzenecarbonitrile, 2-amino-4-methylbenzonitril, 6-cyano-m-toluidine, pubchem4661, 4-methylanthranilonitrile
2801276
2-amino-4-methylbenzonitrile
CC1=CC=C(C#N)C(N)=C1
This item is not returnable. View return policy
26830-96-6
C8H8N2
132.17
MFCD00173706
LGNVAEIITHYWCG-UHFFFAOYSA-N
2-cyano-5-methylaniline, 3-amino-4-cyanotoluene, 2-amino-4-methyl-benzonitrile, 4-methyl-2-aminobenzonitrile, benzonitrile, 2-amino-4-methyl, 2-amino-4-methylbenzenecarbonitrile, 2-amino-4-methylbenzonitril, 6-cyano-m-toluidine, pubchem4661, 4-methylanthranilonitrile
2801276
2-amino-4-methylbenzonitrile
CC1=CC=C(C#N)C(N)=C1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 132.17
Percent Purity 97%
Chemical Name or Material 2-Amino-4-methylbenzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'