missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methoxybenzothiazole, 97%, Thermo Scientific Chemicals

Catalog Number 10490514
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5464-79-9
C8H8N2OS
180.23
MFCD00005792
YEBCRAVYUWNFQT-UHFFFAOYSA-N
2-amino-4-methoxybenzothiazole, 4-methoxy-2-aminobenzothiazole, 2-benzothiazolamine, 4-methoxy, 4-methoxybenzo d thiazol-2-amine, 2-amino-4-methoxy-benzothiazole, benzothiazole, 2-amino-4-methoxy, 4-methoxy-benzothiazol-2-ylamine, 4-methoxybenzothiazol-2-ylamine, 4-methoxybenzothiazole-2-ylamine, 2-benzothiazolamine, 4-methoxy-9ci
21622
4-methoxy-1,3-benzothiazol-2-amine
COC1=C2C(=CC=C1)SC(=N2)N
This item is not returnable. View return policy
5464-79-9
C8H8N2OS
180.23
MFCD00005792
YEBCRAVYUWNFQT-UHFFFAOYSA-N
2-amino-4-methoxybenzothiazole, 4-methoxy-2-aminobenzothiazole, 2-benzothiazolamine, 4-methoxy, 4-methoxybenzo d thiazol-2-amine, 2-amino-4-methoxy-benzothiazole, benzothiazole, 2-amino-4-methoxy, 4-methoxy-benzothiazol-2-ylamine, 4-methoxybenzothiazol-2-ylamine, 4-methoxybenzothiazole-2-ylamine, 2-benzothiazolamine, 4-methoxy-9ci
21622
4-methoxy-1,3-benzothiazol-2-amine
COC1=C2C(=CC=C1)SC(=N2)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 153.0°C to 155.0°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 180.23
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-methoxybenzothiazole
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-4-methoxybenzothiazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'