missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Amino-4-methoxybenzothiazole, 97%

Brand:  Alfa Aesar™ L11358.06

Code : NEW

Additional Details : CAS Number : 5464-79-9 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11307736

  • £88.40 / 5g


Estimated Shipment date
from Supplier 14-05-2018
Add to basket

Specifications

Chemical Identifiers

2-Amino-4-methoxybenzothiazole
C8H8N2OS
MFCD00005792
5464-79-9
180.23
141363

Specifications

2-Amino-4-methoxybenzothiazole
97%
153° to 159°C
MFCD00005792
141363
5464-79-9
C8H8N2OS
180.23
5g
Videos
SDS