missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-hydroxy-5,6-dimethylpyrimidine, 97%, Thermo Scientific™

Catalog Number p-7058892
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
3977-23-9
C6H9N3O
139.158
MFCD00023258
APWRLAZEMYLHKZ-UHFFFAOYSA-N
2-amino-5,6-dimethylpyrimidin-4-ol, 2-amino-5,6-dimethyl-4-hydroxypyrimidine, 2-amino-5,6-dimethylpyrimidin-4 3h-one, 2-amino-5,6-dimethyl-4-pyrimidinol, 2-amino-5,6-dimethyl-4 3h-pyrimidinone, 2-amino-5,6-dimethylpyrimidin-4 1h-one, 2-amino-5,6-dimethyl-3h-pyrimidin-4-one, aminodimethylpyrimidinol, acmc-1cpts, 4 1h-pyrimidinone, 2-amino-5,6-dimethyl
77600
2-amino-5,6-dimethyl-1H-pyrimidin-4-one
CC1=C(NC(=NC1=O)N)C
This item is not returnable. View return policy
3977-23-9
C6H9N3O
139.158
MFCD00023258
APWRLAZEMYLHKZ-UHFFFAOYSA-N
2-amino-5,6-dimethylpyrimidin-4-ol, 2-amino-5,6-dimethyl-4-hydroxypyrimidine, 2-amino-5,6-dimethylpyrimidin-4 3h-one, 2-amino-5,6-dimethyl-4-pyrimidinol, 2-amino-5,6-dimethyl-4 3h-pyrimidinone, 2-amino-5,6-dimethylpyrimidin-4 1h-one, 2-amino-5,6-dimethyl-3h-pyrimidin-4-one, aminodimethylpyrimidinol, acmc-1cpts, 4 1h-pyrimidinone, 2-amino-5,6-dimethyl
77600
2-amino-5,6-dimethyl-1H-pyrimidin-4-one
CC1=C(NC(=NC1=O)N)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 316°C to 318°C (decomposition)
Color White
Quantity 1 g
Formula Weight 139.16
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Amino-4-hydroxy-5,6-dimethylpyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'