missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-fluorobenzoic acid, 96%, Thermo Scientific Chemicals

Catalog Number 10461101
Click to view available options
Quantity:
2.5 g
10 g
This item is not returnable. View return policy
446-32-2
C7H6FNO2
155.13
MFCD00075553
LGPVTNAJFDUWLF-UHFFFAOYSA-N
4-fluoroanthranilic acid, 2-amino-4-fluoro-benzoic acid, benzoic acid, 2-amino-4-fluoro, 2-amino-4-fluoro benzoic acid, 2-amino-4-fluorobenzoicacid, fa0, pubchem1323, amino-4-fluorobenzoic acid, acmc-209jz7, 2-amino-4fluorobenzoic acid
2724967
2-amino-4-fluorobenzoic acid
C1=CC(=C(C=C1F)N)C(=O)O
This item is not returnable. View return policy
446-32-2
C7H6FNO2
155.13
MFCD00075553
LGPVTNAJFDUWLF-UHFFFAOYSA-N
4-fluoroanthranilic acid, 2-amino-4-fluoro-benzoic acid, benzoic acid, 2-amino-4-fluoro, 2-amino-4-fluoro benzoic acid, 2-amino-4-fluorobenzoicacid, fa0, pubchem1323, amino-4-fluorobenzoic acid, acmc-209jz7, 2-amino-4fluorobenzoic acid
2724967
2-amino-4-fluorobenzoic acid
C1=CC(=C(C=C1F)N)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Melting Point 188.0°C to 196.0°C
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Linear Formula H2NC6H3(F)COOH
Quantity 2.5 g
Formula Weight 155.13
Percent Purity 96%
Chemical Name or Material 2-Amino-4-fluorobenzoic acid
compliance-icons
Product Identifier
  • 2-Amino-4-fluorobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'