missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Amino-4-ethylaminothiazole dihydrochloride, 97%

Brand:  Alfa Aesar™ H63633.03

Code : NEW

Additional Details : CAS Number : 89364-16-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15460429

  • £143.10 / 1g


Estimated Shipment date
from Supplier 04-05-2018
Add to basket

Specifications

Chemical Identifiers

2-Amino-4-ethylaminothiazole dihydrochloride
89364-16-9
216.13
MFCD12963986

Specifications

2-Amino-4-ethylaminothiazole dihydrochloride
Solid
1g
MFCD12963986
89364-16-9
97%
216.13
Videos
SDS