Alfa Aesar™ 2-Amino-4-chlorophenol, 98%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: A10135.22

2-AMINO-4-CHLOROPHENOL, 97%,100G

UNSPSC: 12352104

Code: NEW

Additional Details:
Additional Details: CAS Number: 95-85-2 Weight: 0.01000kg


Quantity 100g

Product Code. 11489392

Quantity Price
1 £ 11.129999999999999 / 100g
Estimated Shipment date
from Supplier 30-01-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 2-Amino-4-chlorophenol
CAS 95-85-2
Molecular Formula C6H6ClNO
Formula Weight 143.57
MDL Number MFCD00007694
UN Number UN2673
Beilstein 774859
Synonym 5-Chloro-2-hydroxyaniline

Specification

Formula Weight 143.57
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2-Amino-4-chlorophenol
TSCA TSCA
Melting Point 138° to 142°C
Quantity 100g
Assay 98%
Molecular Formula C6H6ClNO
MDL Number MFCD00007694
CAS 95-85-2
UN Number UN2673
Beilstein 774859
Synonym 5-Chloro-2-hydroxyaniline
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 2-AMINO-4-CHLOROPHENOL
RTECS Number SJ5700000
Safety and Handling

Warning

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.