missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chlorobenzothiazole, 97%, Thermo Scientific™

Catalog Number 10647512
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
19952-47-7
MFCD00005791
OEQQFQXMCPMEIH-UHFFFAOYSA-N
2-amino-4-chlorobenzothiazole, 2-benzothiazolamine, 4-chloro, 4-chlorobenzo d thiazol-2-amine, 4-chlorobenzothiazol-2-ylamine, benzothiazole, 2-amino-4-chloro, 4-chloro-2-benzothiazolamine, 4-chloro-2-aminobenzothiazole, 4-chloro-benzothiazol-2-ylamine, 4-chlorobenzothiazole-2-ylamine, zlchem 166
29872
4-chloro-1,3-benzothiazol-2-amine
C1=CC2=C(C(=C1)Cl)N=C(S2)N
This item is not returnable. View return policy
19952-47-7
MFCD00005791
OEQQFQXMCPMEIH-UHFFFAOYSA-N
2-amino-4-chlorobenzothiazole, 2-benzothiazolamine, 4-chloro, 4-chlorobenzo d thiazol-2-amine, 4-chlorobenzothiazol-2-ylamine, benzothiazole, 2-amino-4-chloro, 4-chloro-2-benzothiazolamine, 4-chloro-2-aminobenzothiazole, 4-chloro-benzothiazol-2-ylamine, 4-chlorobenzothiazole-2-ylamine, zlchem 166
29872
4-chloro-1,3-benzothiazol-2-amine
C1=CC2=C(C(=C1)Cl)N=C(S2)N

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 203.0°C to 208.0°C
Color Beige to White or Gray
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C7H5ClN2S
Quantity 5 g
Molecular Weight (g/mol) 184.64
Formula Weight 184.64
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-chlorobenzothiazole, 97%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'