missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chlorobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-3777234
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
89-77-0
C7H6ClNO2
171.58
MFCD00007778
JYYLQSCZISREGY-UHFFFAOYSA-N
4-chloroanthranilic acid, benzoic acid, 2-amino-4-chloro, 4-chloro-2-aminobenzoic acid, anthranilic acid, 4-chloro, 4-chloroanthranil acid, 4-chloro anthranilic acid, 2-carboxy-5-chloroaniline, unii-g55k85r12h, 2-amino-4-chloro-benzoic acid, 2,4-acba
66646
2-amino-4-chlorobenzoic acid
NC1=CC(Cl)=CC=C1C(O)=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
89-77-0
C7H6ClNO2
171.58
MFCD00007778
JYYLQSCZISREGY-UHFFFAOYSA-N
4-chloroanthranilic acid, benzoic acid, 2-amino-4-chloro, 4-chloro-2-aminobenzoic acid, anthranilic acid, 4-chloro, 4-chloroanthranil acid, 4-chloro anthranilic acid, 2-carboxy-5-chloroaniline, unii-g55k85r12h, 2-amino-4-chloro-benzoic acid, 2,4-acba
66646
2-amino-4-chlorobenzoic acid
NC1=CC(Cl)=CC=C1C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 231°C to 235°C
Color Beige to Brown
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Linear Formula H2NC6H3(Cl)CO2H
Quantity 25 g
Beilstein 14, 365
Solubility Information Solubility in water: slightly soluble
Formula Weight 171.58
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Amino-4-chlorobenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-4-chlorobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'