missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Amino-4-chlorobenzoic acid, 98%

Brand:  Alfa Aesar™ A10201.06

Code : NEW

Additional Details : CAS Number : 89-77-0 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11369689

  • £14.67 / 5g

In stock, ready to ship: 2

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

89-77-0
171.58
JYYLQSCZISREGY-UHFFFAOYSA-N
66646
NC1=CC(Cl)=CC=C1C(O)=O
C7H6ClNO2
MFCD00007778
4-chloroanthranilic acid, benzoic acid, 2-amino-4-chloro, 4-chloro-2-aminobenzoic acid, anthranilic acid, 4-chloro, 4-chloroanthranil acid, 4-chloro anthranilic acid, 2-carboxy-5-chloroaniline, unii-g55k85r12h, 2-amino-4-chloro-benzoic acid, 2,4-acba
2-amino-4-chlorobenzoic acid

Specifications

2-Amino-4-chlorobenzoic acid
5g
C7H6ClNO2
743349
JYYLQSCZISREGY-UHFFFAOYSA-N
2-amino-4-chlorobenzoic acid
66646
98%
~232°C (decomposition)
89-77-0
MFCD00007778
4-chloroanthranilic acid, benzoic acid, 2-amino-4-chloro, 4-chloro-2-aminobenzoic acid, anthranilic acid, 4-chloro, 4-chloroanthranil acid, 4-chloro anthranilic acid, 2-carboxy-5-chloroaniline, unii-g55k85r12h, 2-amino-4-chloro-benzoic acid, 2,4-acba
NC1=CC(Cl)=CC=C1C(O)=O
171.58
171.58
Videos
SDS Documents