missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Amino-4-chloro-6-methylpyrimidine, 98%

Brand:  Alfa Aesar™ L06896.14

Code : NEW

Additional Details : CAS Number : 5600-21-5 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11307046

  • £53.50 / 25g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Description

Description

Applications
2-Amino-4-chloro-6-methylpyrimidine is a nitrification inhibitor. It was used to study the influence of chlorine substitution in pyrimidine ring on proton donor ability of amino group in 2-aminopyrimidine.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

2-Amino-4-chloro-6-methylpyrimidine
C5H6ClN3
MFCD00006091
5600-21-5
143.58
114297

Specifications

2-Amino-4-chloro-6-methylpyrimidine
181° to 186°C
98%
143.58
114297
5600-21-5
25g
C5H6ClN3
MFCD00006091
Videos
SDS