missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chloro-6-methoxypyrimidine, 95%, ACROS Organics™

Brand:  Acros Organics 347090250

Code : 29

Additional Details : CAS Number : 5734-64-5 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10596641

  • £105.20 / 25g


Estimated Shipment
15-06-2018
Add to basket


Specifications

Chemical Identifiers

2-Amino-4-chloro-6-methoxypyrimidine
5734-64-5
100.0
95%
159.57
95%
94.0
≥94%
C5H6ClN3O
MFCD00233535

Specifications

2-Amino-4-chloro-6-methoxypyrimidine
Glass bottle
Authentic
100.0
Light yellow
Crystalline Powder
95%
159.57
95%
5734-64-5
94.0
169° to 176°C
25g
≥94%
C5H6ClN3O
MFCD00233535
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Amino-4-chloro-6-methoxypyrimidine

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

SDS