missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chloro-6-methoxypyrimidine, 95%, Thermo Scientific™

Catalog Number p-54937
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5734-64-5
C5H6ClN3O
159.57
MFCD00233535
VFEYBTFCBZMBAU-UHFFFAOYSA-N
2-amino-4-chloro-6-methoxypyrimidine, 2-pyrimidinamine, 4-chloro-6-methoxy, 2-amino-6-chloro-4-methoxypyrimidine, pyrimidine, 2-amino-4-chloro-6-methoxy, 2-amino-4-methoxy-6-chloro pyrimidine, 4-chloro-6-methoxy-2-pyrimidinamine, 2-amino-4-chloro-6-methoxylpyrimidine, 2-amino-4-chloro-6-methoxy pyrimidine, 2-amino-4-chloro-6-methoxy-pyrimidine, 4-chloro-6-methoxy-pyrimidin-2-ylamine
79811
4-chloro-6-methoxypyrimidin-2-amine
COC1=CC(Cl)=NC(N)=N1
This item is not returnable. View return policy
5734-64-5
C5H6ClN3O
159.57
MFCD00233535
VFEYBTFCBZMBAU-UHFFFAOYSA-N
2-amino-4-chloro-6-methoxypyrimidine, 2-pyrimidinamine, 4-chloro-6-methoxy, 2-amino-6-chloro-4-methoxypyrimidine, pyrimidine, 2-amino-4-chloro-6-methoxy, 2-amino-4-methoxy-6-chloro pyrimidine, 4-chloro-6-methoxy-2-pyrimidinamine, 2-amino-4-chloro-6-methoxylpyrimidine, 2-amino-4-chloro-6-methoxy pyrimidine, 2-amino-4-chloro-6-methoxy-pyrimidine, 4-chloro-6-methoxy-pyrimidin-2-ylamine
79811
4-chloro-6-methoxypyrimidin-2-amine
COC1=CC(Cl)=NC(N)=N1
CAS Min % 94.0
CAS Max % 100.0
Melting Point 169°C to 176°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 5 g
Formula Weight 159.57
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-chloro-6-methoxypyrimidine, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-4-chloro-6-methoxypyrimidine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'