missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chloro-5,6-dimethylpyrimidine, 97%, Thermo Scientific Chemicals

Catalog Number p-7036287
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
14394-61-7
C6H8ClN3
157.601
MFCD00462111
RRMVDLLCVGZDMH-UHFFFAOYSA-N
248655
4-chloro-5,6-dimethylpyrimidin-2-amine
CC1=C(N=C(N=C1Cl)N)C
This item is not returnable. View return policy
14394-61-7
C6H8ClN3
157.601
MFCD00462111
RRMVDLLCVGZDMH-UHFFFAOYSA-N
248655
4-chloro-5,6-dimethylpyrimidin-2-amine
CC1=C(N=C(N=C1Cl)N)C

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in Chloroform and Ethanol

Notes
Store at -20°C. Keep away from light and heat.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Synonym 4-chloro-5,6-dimethyl-pyrimidin-2-ylamine, 2-amino-4-chloro-5,6-dimethylpyrimidine, 2-pyrimidinamine, 4-chloro-5,6-dimethyl, 2-amino-4-chloro-5,6-dimethyl-pyrimidine, 6-chloro-4,5-dimethylpyrimidine-2-ylamine, 2-pyrimidinamine,4-chloro-5,6-dimethyl
Solubility Information Soluble in Chloroform and Ethanol
Formula Weight 157.6
Percent Purity 97%
Chemical Name or Material 2-Amino-4-chloro-5,6-dimethylpyrimidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'