missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-bromopyrimidine, 98%, Thermo Scientific Chemicals

Catalog Number p-7055710
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
343926-69-2
C4H4BrN3
174.001
MFCD09999168
MINURGGJJKMQQQ-UHFFFAOYSA-N
2-amino-4-bromopyrimidine, 4-bromo-2-pyrimidinamine, 2-pyrimidinamine,4-bromo, 4-bromanylpyrimidin-2-amine, 4-bromopyrimidine-2-ylamine, 2-amino-6-bromopyrimidine, 4-bromo-2-aminopyrimidine, 2-pyrimidinamine,4-bromo-9ci, 2-pyrimidinamine, 4-bromo, 4-bromo-pyrimidin-2-ylamine
12803496
4-bromopyrimidin-2-amine
C1=CN=C(N=C1Br)N
This item is not returnable. View return policy
343926-69-2
C4H4BrN3
174.001
MFCD09999168
MINURGGJJKMQQQ-UHFFFAOYSA-N
2-amino-4-bromopyrimidine, 4-bromo-2-pyrimidinamine, 2-pyrimidinamine,4-bromo, 4-bromanylpyrimidin-2-amine, 4-bromopyrimidine-2-ylamine, 2-amino-6-bromopyrimidine, 4-bromo-2-aminopyrimidine, 2-pyrimidinamine,4-bromo-9ci, 2-pyrimidinamine, 4-bromo, 4-bromo-pyrimidin-2-ylamine
12803496
4-bromopyrimidin-2-amine
C1=CN=C(N=C1Br)N

It is used as a pharmaceutical intermediate. It is also an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate. It is also an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
Beilstein 111375
Solubility Information Slightly soluble in water.
Formula Weight 174
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Amino-4-bromopyrimidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'