missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4'-bromoacetophenone hydrochloride, 97%, Thermo Scientific™

Catalog Number p-3771017
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5467-72-1
C8H8BrNO·ClH
250.52
MFCD00051998
ROAVTVXTYFSQEA-UHFFFAOYSA-N
4-bromophenacylamine hydrochloride, 2-amino-4'-bromoacetophenone hydrochloride, 2-amino-1-4-bromophenyl ethanone hydrochloride, 2-amino-4'-bromoacetophenone hcl, 2-amino-4-bromoacetophenone hydrochloride, 2-amino-1-4-bromophenyl ethan-1-one hydrochloride, a-amino-4'-bromoacetophenone, ethanone, 2-amino-1-4-bromophenyl-, hydrochloride, 2-amino-1-4-bromophenyl ethan-1-one, ethanone, 2-amino-1-4-bromophenyl
2798216
2-amino-1-(4-bromophenyl)ethanone;hydrochloride
C1=CC(=CC=C1C(=O)CN)Br.Cl
This item is not returnable. View return policy
5467-72-1
C8H8BrNO·ClH
250.52
MFCD00051998
ROAVTVXTYFSQEA-UHFFFAOYSA-N
4-bromophenacylamine hydrochloride, 2-amino-4'-bromoacetophenone hydrochloride, 2-amino-1-4-bromophenyl ethanone hydrochloride, 2-amino-4'-bromoacetophenone hcl, 2-amino-4-bromoacetophenone hydrochloride, 2-amino-1-4-bromophenyl ethan-1-one hydrochloride, a-amino-4'-bromoacetophenone, ethanone, 2-amino-1-4-bromophenyl-, hydrochloride, 2-amino-1-4-bromophenyl ethan-1-one, ethanone, 2-amino-1-4-bromophenyl
2798216
2-amino-1-(4-bromophenyl)ethanone;hydrochloride
C1=CC(=CC=C1C(=O)CN)Br.Cl
CAS Min % 96.0
CAS Max % 100.0
Melting Point 280°C to 285°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula BrC6H4C(O)CH2NH2·HCl
Quantity 1 g
Solubility Information Solubility in water: soluble
Formula Weight 250.52
Percent Purity 97%
Chemical Name or Material 2-Amino-4′-bromoacetophenone hydrochloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'