missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,6-dimethylpyridine, 99%, ACROS Organics™

Brand:  Acros Organics 103580250

Code : 29

Additional Details : CAS Number : 5407-87-4 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 11947031

  • £130.50 / 25g


Estimated Shipment
18-06-2018
Add to basket


Specifications

Chemical Identifiers

2-Amino-4, 6-dimethylpyridine
98.5
99%
C7H10N2
122.17
5407-87-4
100.0
99%
(CH3)2(C5H2N)NH2
6-Amino-2, 4-lutidine

Specifications

2-Amino-4, 6-dimethylpyridine
98.5
Authentic
Solubility in water: 5.5 g/100mL (20 c).
65.0° to 69.5°C
Crystalline Powder
25g
121°C
(CH3)2(C5H2N)NH2
6-Amino-2, 4-lutidine
5407-87-4
100.0
Glass bottle
White to light brown
235.0°C
99%
99%
C7H10N2
122.17
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Amino-4,6-dimethylpyridine, 99%

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS