missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(3-pyridyl)thiazole, 97%, Thermo Scientific™

Catalog Number p-7053005
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
30235-27-9
C8H7N3S
177.225
MFCD00218013
XOHZQGAYUHOJPR-UHFFFAOYSA-N
4-pyridin-3-yl thiazol-2-amine, 2-amino-4-3-pyridyl thiazole, 4-pyridin-3-yl-thiazol-2-ylamine, 4-pyridin-3-yl-1,3-thiazol-2-amine, 4-3-pyridinyl-2-thiazolamine, 2-thiazolamine, 4-3-pyridinyl, ccris 6699, 2-amino-4-pyridin-3-yl-1,3-thiazole, 4-pyridine-3-yl-thiazol-2-ylamine, 4-3-pyridinyl-1,3-thiazol-2-ylamine
154893
4-pyridin-3-yl-1,3-thiazol-2-amine
C1=CC(=CN=C1)C2=CSC(=N2)N
This item is not returnable. View return policy
30235-27-9
C8H7N3S
177.225
MFCD00218013
XOHZQGAYUHOJPR-UHFFFAOYSA-N
4-pyridin-3-yl thiazol-2-amine, 2-amino-4-3-pyridyl thiazole, 4-pyridin-3-yl-thiazol-2-ylamine, 4-pyridin-3-yl-1,3-thiazol-2-amine, 4-3-pyridinyl-2-thiazolamine, 2-thiazolamine, 4-3-pyridinyl, ccris 6699, 2-amino-4-pyridin-3-yl-1,3-thiazole, 4-pyridine-3-yl-thiazol-2-ylamine, 4-3-pyridinyl-1,3-thiazol-2-ylamine
154893
4-pyridin-3-yl-1,3-thiazol-2-amine
C1=CC(=CN=C1)C2=CSC(=N2)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as a pharmaceutical intermediate.

Notes
Store at room temperature. Incompatible with strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY
Color Brown
Quantity 2 g
Formula Weight 177.22
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Amino-4-(3-pyridyl)thiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'