missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(2-furyl)thiazole, 97%, Thermo Scientific™

Catalog Number p-7053048
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
28989-52-8
C7H6N2OS
166.20
MFCD01167261
CGJGALUSBPYALD-UHFFFAOYSA-N
4-furan-2-yl thiazol-2-amine, 4-furan-2-yl-1,3-thiazol-2-amine, 4-2-furyl-1,3-thiazol-2-amine, 2-amino-4-2-furyl thiazole, 2-thiazolamine, 4-2-furanyl, 4-furan-2-yl-thiazol-2-ylamine, 4-2-furyl thiazole-2-amine, 4-2-furanyl-2-thiazolamine, cgjgalusbpyald-uhfffaoysa, 2-thiazol amine,4-2-furanyl
727834
4-(furan-2-yl)-1,3-thiazol-2-amine
NC1=NC(=CS1)C1=CC=CO1
This item is not returnable. View return policy
28989-52-8
C7H6N2OS
166.20
MFCD01167261
CGJGALUSBPYALD-UHFFFAOYSA-N
4-furan-2-yl thiazol-2-amine, 4-furan-2-yl-1,3-thiazol-2-amine, 4-2-furyl-1,3-thiazol-2-amine, 2-amino-4-2-furyl thiazole, 2-thiazolamine, 4-2-furanyl, 4-furan-2-yl-thiazol-2-ylamine, 4-2-furyl thiazole-2-amine, 4-2-furanyl-2-thiazolamine, cgjgalusbpyald-uhfffaoysa, 2-thiazol amine,4-2-furanyl
727834
4-(furan-2-yl)-1,3-thiazol-2-amine
NC1=NC(=CS1)C1=CC=CO1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Brown
Quantity 5 g
Formula Weight 166.2
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Amino-4-(2-furyl)thiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'