missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Amino-4-(2-chloro-3-pyridyl)pyrimidine, 97%

Brand:  Alfa Aesar™ H54957.03

Code : NEW

Additional Details : CAS Number : 870221-49-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15401098

  • £315.00 / 1g

Estimated Shipment: 26-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Applications
It is used as primary and secondary intermediate.

Notes
Incompatible with strong oxidizing agents, bases, acids. Keep in room temperature.
Specifications

Chemical Identifiers

870221-49-1
206.633
DKCVVHMYFZBDAK-UHFFFAOYSA-N
53396335
C1=CC(=C(N=C1)Cl)C2=NC(=NC=C2)N
C9H7ClN4
MFCD18157687
4-2-chloropyridin-3-yl pyrimidin-2-amine, chloropyridinylpyrimidinamine, 2-pyrimidinamine,4-2-chloro-3-pyridinyl, 2-pyrimidinamine, 4-2-chloro-3-pyridinyl
4-(2-chloropyridin-3-yl)pyrimidin-2-amine

Specifications

2-Amino-4-(2-chloro-3-pyridyl)pyrimidine
870221-49-1
MFCD18157687
4-2-chloropyridin-3-yl pyrimidin-2-amine, chloropyridinylpyrimidinamine, 2-pyrimidinamine,4-2-chloro-3-pyridinyl, 2-pyrimidinamine, 4-2-chloro-3-pyridinyl
C1=CC(=C(N=C1)Cl)C2=NC(=NC=C2)N
206.633
206.63
1g
C9H7ClN4
14484335
DKCVVHMYFZBDAK-UHFFFAOYSA-N
4-(2-chloropyridin-3-yl)pyrimidin-2-amine
53396335
97%
Videos
SDS Documents
Certificates