missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Amino-4-(2,4-difluorophenyl)thiazole, 97%

£35.46 - £91.52

Chemical Identifiers

CAS 105512-80-9
Molecular Formula C9H6F2N2S
Molecular Weight (g/mol) 212.22
MDL Number MFCD00472891
InChI Key HKUPOLYVZQLXKP-UHFFFAOYSA-N
Synonym 4-2,4-difluorophenyl-1,3-thiazol-2-amine, 2-amino-4-2,4-difluorophenyl thiazole, 4-2,4-difluorophenyl thiazol-2-amine, 2-amino-4-2,4-difluorophenyl-1,3-thiazole, 4-2,4-difluorophenyl thiazol-2-ylamine, 4-2,4-difluoro-phenyl-thiazol-2-ylamine, 4-2,4-difluorophenyl-1,3-thiazole-2-ylamine, acmc-1bro0, cbmicro_034355, 2-thiazolamine,4-2,4-difluorophenyl
PubChem CID 611370
IUPAC Name 4-(2,4-difluorophenyl)-1,3-thiazol-2-amine
SMILES NC1=NC(=CS1)C1=C(F)C=C(F)C=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15454597
SDS View Documents
Alfa Aesar™
H31948.ME
500mg
£35.46
500mg
View alternative products
Save Up to 
»
Estimated Shipment: 09-08-2021
Please to purchase this item. Need a web account? Register with us today!
15444597
SDS View Documents
Alfa Aesar™
H31948.04
2g
£91.52
2g
View alternative products
Save Up to 
»
Estimated Shipment: 02-08-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

105512-80-9
212.22
HKUPOLYVZQLXKP-UHFFFAOYSA-N
611370
NC1=NC(=CS1)C1=C(F)C=C(F)C=C1
C9H6F2N2S
MFCD00472891
4-2,4-difluorophenyl-1,3-thiazol-2-amine, 2-amino-4-2,4-difluorophenyl thiazole, 4-2,4-difluorophenyl thiazol-2-amine, 2-amino-4-2,4-difluorophenyl-1,3-thiazole, 4-2,4-difluorophenyl thiazol-2-ylamine, 4-2,4-difluoro-phenyl-thiazol-2-ylamine, 4-2,4-difluorophenyl-1,3-thiazole-2-ylamine, acmc-1bro0, cbmicro_034355, 2-thiazolamine,4-2,4-difluorophenyl
4-(2,4-difluorophenyl)-1,3-thiazol-2-amine

Specifications

2-Amino-4-(2,4-difluorophenyl)thiazole
105512-80-9
MFCD00472891
HKUPOLYVZQLXKP-UHFFFAOYSA-N
4-(2,4-difluorophenyl)-1,3-thiazol-2-amine
611370
97%
500mg
C9H6F2N2S
4-2,4-difluorophenyl-1,3-thiazol-2-amine, 2-amino-4-2,4-difluorophenyl thiazole, 4-2,4-difluorophenyl thiazol-2-amine, 2-amino-4-2,4-difluorophenyl-1,3-thiazole, 4-2,4-difluorophenyl thiazol-2-ylamine, 4-2,4-difluoro-phenyl-thiazol-2-ylamine, 4-2,4-difluorophenyl-1,3-thiazole-2-ylamine, acmc-1bro0, cbmicro_034355, 2-thiazolamine,4-2,4-difluorophenyl
NC1=NC(=CS1)C1=C(F)C=C(F)C=C1
212.22
212.22
SDS
Product Certifications