missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-picoline 96%, ACROS Organics™

Brand:  Acros Organics 104385000

UNSPSC : 12350000

Code : 29

Additional Details : CAS Number : 1603-40-3 Weight : 0.55000kg Transport : UN number : 2811 Chem class : 1 Pack group : III


Quantity 500g
Packaging Glass bottle

 View more versions of this product

Product Code. 10714211

  • £91.50 / 500g


Estimated Shipment
16-10-2017
Add to basket


Specifications

Chemical Identifiers

2-Amino-3-picoline
1603-40-3
100.0
96%
108.14
2-Amino-3-methylpyridine
96%
95.0
96%
C6H8N2
MFCD00006320

Specifications

2-Amino-3-picoline
1603-40-3
95.0
Glass bottle
Solubility in water: freely soluble.
Clear yellow
221.0° to 222.0°C
500g
96%
C6H8N2
MFCD00006320
2-Amino-3-methylpyridine
96%
Authentic
100.0
1.5813 to 1.5833
0.8% max.
32.0° to 34.0°C
Liquid After Melting
111°C
96%
108.14
22, V,9, 272
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Amino-3-picoline

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /A§
  • P302+P350-IF ON SKIN: Gently wash with plenty of soap and water.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/A§/if you feel unwell.

Documentation

Technical Documents