missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-nitropyridine 99%, ACROS Organics™

Brand:  Acros Organics 151120050

Code : 29

Additional Details : CAS Number : 4214-75-9 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10776541

  • £21.10 / 5g


EU Stock 1
Add to basket


Specifications

Chemical Identifiers

2-Amino-3-nitropyridine
4214-75-9
100.0
99%
NH2(C5H3N)NO2
MFCD00006314
99%
98.5
≥98.5%
C5H5N3O2
139.11

Specifications

2-Amino-3-nitropyridine
4214-75-9
100.0
Glass bottle
Yellow
≥98.5%
5g
NH2(C5H3N)NO2
MFCD00006314
99%
98.5
Authentic
164° to 168°C
Crystalline Powder and Needles
99%
C5H5N3O2
139.11
22, I, 631
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Amino-3-nitropyridine

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS