missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-nitrobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-4812731
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
606-18-8
C7H6N2O4
182.13
JJPIVRWTAGQTPQ-UHFFFAOYSA-N
2-amino-3-nitrobenzoic acid, 3-nitroanthranilic acid, benzoic acid, 2-amino-3-nitro, 2-amino-3-nitrobenzonic acid, 2-amino-3-nitro-benzoic acid, 2-amino-3-nitro benzoic acid, pubchem2233, amino-3-nitrobenzoic acid, 2-amino-nitro-benzoesaure, 2-carboxy-6-nitroaniline
219633
C1=CC(=C(C(=C1)[N+](=O)[O-])N)C(=O)O
This item is not returnable. View return policy
606-18-8
C7H6N2O4
182.13
JJPIVRWTAGQTPQ-UHFFFAOYSA-N
2-amino-3-nitrobenzoic acid, 3-nitroanthranilic acid, benzoic acid, 2-amino-3-nitro, 2-amino-3-nitrobenzonic acid, 2-amino-3-nitro-benzoic acid, 2-amino-3-nitro benzoic acid, pubchem2233, amino-3-nitrobenzoic acid, 2-amino-nitro-benzoesaure, 2-carboxy-6-nitroaniline
219633
C1=CC(=C(C(=C1)[N+](=O)[O-])N)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 204.0°C to 207.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 182.13
Percent Purity 97%
Chemical Name or Material 2-Amino-3-nitrobenzoic acid
compliance-icons
Product Identifier
  • 2-Amino-3-nitrobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'