missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Amino-3-nitrobenzoic acid, 97%

Brand:  Alfa Aesar™ H26001.03

Code : NEW

Additional Details : CAS Number : 606-18-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15403337

  • £18.59 / 1g

Estimated Shipment: 12-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

606-18-8
182.135
JJPIVRWTAGQTPQ-UHFFFAOYSA-N
219633
C7H6N2O4
MFCD00024261
2-amino-3-nitrobenzoic acid, 3-nitroanthranilic acid, benzoic acid, 2-amino-3-nitro, 2-amino-3-nitrobenzonic acid, 2-amino-3-nitro-benzoic acid, 2-amino-3-nitro benzoic acid, pubchem2233, amino-3-nitrobenzoic acid, 2-amino-nitro-benzoesaure, 2-carboxy-6-nitroaniline
C1=CC(=C(C(=C1)[N+](=O)[O-])N)C(=O)O

Specifications

2-Amino-3-nitrobenzoic acid
1g
C7H6N2O4
2-amino-3-nitrobenzoic acid, 3-nitroanthranilic acid, benzoic acid, 2-amino-3-nitro, 2-amino-3-nitrobenzonic acid, 2-amino-3-nitro-benzoic acid, 2-amino-3-nitro benzoic acid, pubchem2233, amino-3-nitrobenzoic acid, 2-amino-nitro-benzoesaure, 2-carboxy-6-nitroaniline
C1=CC(=C(C(=C1)[N+](=O)[O-])N)C(=O)O
219633
97%
210°C to 212°C
606-18-8
MFCD00024261
JJPIVRWTAGQTPQ-UHFFFAOYSA-N
182.135
182.14
Videos
SDS Documents
Certificates