missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-methoxypyridine, 97%, Thermo Scientific™

Catalog Number p-4812729
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
10201-71-5
C6H8N2O
124.14
MFCD07374874
HNAYRVKSWGSQTP-UHFFFAOYSA-N
2-amino-3-methoxypyridine, 2-pyridinamine, 3-methoxy, 3-methoxy-2-pyridylamine, 3-methoxy-pyridin-2-ylamine, 3-methoxy-2-pyridinamine, acmc-1c0wi, 3-methoxypyridine-2-amine, 3-methoxy-2-aminopyridine, 3-methoxy-pyridin-2-amine, ksc494m4p
9920384
3-methoxypyridin-2-amine
COC1=C(N)N=CC=C1
This item is not returnable. View return policy
10201-71-5
C6H8N2O
124.14
MFCD07374874
HNAYRVKSWGSQTP-UHFFFAOYSA-N
2-amino-3-methoxypyridine, 2-pyridinamine, 3-methoxy, 3-methoxy-2-pyridylamine, 3-methoxy-pyridin-2-ylamine, 3-methoxy-2-pyridinamine, acmc-1c0wi, 3-methoxypyridine-2-amine, 3-methoxy-2-aminopyridine, 3-methoxy-pyridin-2-amine, ksc494m4p
9920384
3-methoxypyridin-2-amine
COC1=C(N)N=CC=C1
Melting Point 75°C to 77°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 124.14
Percent Purity 97%
Chemical Name or Material 2-Amino-3-methoxypyridine
compliance-icons
Product Identifier
  • 2-Amino-3-methoxypyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'