missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-3-cyano-4,5-di(fur-2-yl)furan, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
IUPAC Name | 5'-amino-2'-(furan-2-yl)-[2,3'-bifuran]-4'-carbonitrile |
Formula Weight | 240.22 |
Percent Purity | 97% |
Chemical Name or Material | 2-Amino-3-cyano-4, 5-di(fur-2-yl)furan |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'