missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-chloropyridine, 97%, Thermo Scientific™

Catalog Number 15209866
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
39620-04-7
C5H5ClN2
128.56
RZJPBQGRCNJYBU-UHFFFAOYSA-N
2-amino-3-chloropyridine, 3-chloro-pyridin-2-ylamine, 2-amino-3-chloro pyridine, 3-chloro-2-pyridinamine, 3-chloro-2-pyridinylamine, 2-pyridinamine, 3-chloro, 3-chloro-2-pyridylamine, 2-amino-3-chloro-pyridine, 3-chloranylpyridin-2-amine, o-aminochloropyridine
693267
3-chloropyridin-2-amine
C1=CC(=C(N=C1)N)Cl
This item is not returnable. View return policy
39620-04-7
C5H5ClN2
128.56
RZJPBQGRCNJYBU-UHFFFAOYSA-N
2-amino-3-chloropyridine, 3-chloro-pyridin-2-ylamine, 2-amino-3-chloro pyridine, 3-chloro-2-pyridinamine, 3-chloro-2-pyridinylamine, 2-pyridinamine, 3-chloro, 3-chloro-2-pyridylamine, 2-amino-3-chloro-pyridine, 3-chloranylpyridin-2-amine, o-aminochloropyridine
693267
3-chloropyridin-2-amine
C1=CC(=C(N=C1)N)Cl
Melting Point 58.0°C to 60.0°C
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 128.56
Percent Purity 97%
Chemical Name or Material 2-Amino-3-chloropyridine
compliance-icons
Product Identifier
  • 2-Amino-3-chloropyridine, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'