missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Amino-3-bromopyridine-5-sulfonic acid, 95%

Brand:  Alfa Aesar™ H61772.03

Code : NEW

Additional Details : CAS Number : 247582-62-3 Weight : 0.00100kg Transport : UN number : 3261 Chem class : 8 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15418348

  • £222.49 / 1g


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Specifications

Chemical Identifiers

2-Amino-3-bromopyridine-5-sulfonic acid
253.07
UN3261
247582-62-3
MFCD03085772
6-Amino-5-bromopyridine-3-sulfonic acid

Specifications

2-Amino-3-bromopyridine-5-sulfonic acid
Solid
Pale brown
1g
UN3261
247582-62-3
95%
253.07
MFCD03085772
6-Amino-5-bromopyridine-3-sulfonic acid
Videos
SDS