missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-3-bromobenzoic acid, 97%, Thermo Scientific Chemicals
30.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
Used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as pharmaceutical intermediates.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Used as pharmaceutical intermediates.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
Melting Point | 174°C to 178°C |
Color | White to Yellow |
Flash Point | 161°C (322°F) |
Quantity | 1 g |
Beilstein | 2936928 |
Formula Weight | 216.03 |
Percent Purity | 97% |
Physical Form | Solid |
Chemical Name or Material | 2-Amino-3-bromobenzoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'