missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-bromo-5-nitropyridine, 98%, Thermo Scientific™

Catalog Number p-3777222
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
15862-31-4
C5H4BrN3O2
218.02
MFCD00955628
OFXNHXMPRZDIDM-UHFFFAOYSA-N
2-amino-3-bromo-5-nitropyridine, 3-bromo-5-nitro-2-pyridylamine, 2-pyridinamine, 3-bromo-5-nitro, 3-bromo-5-nitro-pyridin-2-ylamine, 3-bromo-5-nitro-pyridin-2-amine, 5-nitro-3-bromo-2-aminopyridine, pubchem1148, acmc-209dib, ksc496a7l, cbi-bb zero/006111
2734412
3-bromo-5-nitropyridin-2-amine
C1=C(C=NC(=C1Br)N)[N+](=O)[O-]
This item is not returnable. View return policy
15862-31-4
C5H4BrN3O2
218.02
MFCD00955628
OFXNHXMPRZDIDM-UHFFFAOYSA-N
2-amino-3-bromo-5-nitropyridine, 3-bromo-5-nitro-2-pyridylamine, 2-pyridinamine, 3-bromo-5-nitro, 3-bromo-5-nitro-pyridin-2-ylamine, 3-bromo-5-nitro-pyridin-2-amine, 5-nitro-3-bromo-2-aminopyridine, pubchem1148, acmc-209dib, ksc496a7l, cbi-bb zero/006111
2734412
3-bromo-5-nitropyridin-2-amine
C1=C(C=NC(=C1Br)N)[N+](=O)[O-]
CAS Min % 97.5
CAS Max % 100.0
Melting Point 215°C to 219°C
Color Beige to Brown-Orange
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 218.02
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Amino-3-bromo-5-nitropyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-3-bromo-5-nitropyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'