missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-3-bromo-5-methylpyridine, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 55.0°C to 57.0°C |
Color | Beige to Yellow or Brown |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Glass bottle |
Molecular Formula | C6H7BrN2 |
Quantity | 1 g |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in methanol,chloroform |
Show More |
Safety and Handling
Product Identifier
- 2-Amino-3-bromo-5-methylpyridine
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'