missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-benzyloxypyridine, 98.5%, Thermo Scientific™

Catalog Number p-54829
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
24016-03-3
C12H12N2O
200.24
MFCD00006316
NMCBWICNRJLKKM-UHFFFAOYSA-N
2-amino-3-benzyloxypyridine, 3-benzyloxy pyridin-2-amine, 3-benzyloxy-2-pyridylamine, 2-pyridinamine, 3-phenylmethoxy, 2-amino-3-benzyloxy pyridine, 3ip, 3-phenylmethoxy-2-pyridylamine, 3-benzyloxy pyridin-2-amin, 3fty
90334
3-phenylmethoxypyridin-2-amine
NC1=NC=CC=C1OCC1=CC=CC=C1
This item is not returnable. View return policy
24016-03-3
C12H12N2O
200.24
MFCD00006316
NMCBWICNRJLKKM-UHFFFAOYSA-N
2-amino-3-benzyloxypyridine, 3-benzyloxy pyridin-2-amine, 3-benzyloxy-2-pyridylamine, 2-pyridinamine, 3-phenylmethoxy, 2-amino-3-benzyloxy pyridine, 3ip, 3-phenylmethoxy-2-pyridylamine, 3-benzyloxy pyridin-2-amin, 3fty
90334
3-phenylmethoxypyridin-2-amine
NC1=NC=CC=C1OCC1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point 94°C to 98°C
Color Brown to Green
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 200.24
Percent Purity 98.50%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3-benzyloxypyridine, 98.5%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'