missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Amino-3-benzyloxypyrazine, 97%

Brand:  Alfa Aesar™ H63162.06

Code : NEW

Additional Details : CAS Number : 110223-15-9 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15429698

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

2-Amino-3-benzyloxypyrazine
201.22
3-(Benzyloxy)pyrazin-2-amine
110223-15-9
MFCD09838954

Specifications

2-Amino-3-benzyloxypyrazine
Solid
201.22
MFCD09838954
110223-15-9
97%
5g
3-(Benzyloxy)pyrazin-2-amine
Videos
SDS