missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,5-dichloropyridine 97%, ACROS Organics™

Brand:  Acros Organics 159381000

Code : NEW

Additional Details : CAS Number : 4214-74-8 Weight : 0.15000kg


Quantity 100g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10676742

  • £158.00 / 100g


Estimated Shipment
06-02-2018
Add to basket


Specifications

Chemical Identifiers

2-Amino-3, 5-dichloropyridine
4214-74-8
100.0
97%
Cl2(C5H2N)NH2
MFCD00006313
97%
96.0
≥96.0% (HPLC)
C5H4Cl2N2
163.01

Specifications

2-Amino-3, 5-dichloropyridine
4214-74-8
100.0
Plastic bottle
Off-white to pink to beige
Powder or Crystals
100g
C5H4Cl2N2
163.01
22, 430
97%
96.0
Authentic
0.5% Max.
79° to 83°C
≥96.0% (HPLC)
97%
Cl2(C5H2N)NH2
MFCD00006313
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Amino-3,5-dichloropyridine

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.