missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2-methyl-1-propanol, 99%, Thermo Scientific Chemicals

Catalog Number p-3735477
Click to view available options
Quantity:
500 mL
1 L
5 L
Packaging:
Glass bottle
Plastic drum
124-68-5
C4H11NO
89.14
MFCD00008051
CBTVGIZVANVGBH-UHFFFAOYSA-N
2-amino-2-methyl-1-propanol, isobutanol-2-amine, 1-propanol, 2-amino-2-methyl, amp regular, 2-aminoisobutanol, aminomethylpropanol, 2-amino-2-methylpropanol, 2-methyl-2-aminopropanol, 2-aminodimethylethanol, hydroxy-tert-butylamine
11807
2-amino-2-methylpropan-1-ol
CC(C)(N)CO
124-68-5
C4H11NO
89.14
MFCD00008051
CBTVGIZVANVGBH-UHFFFAOYSA-N
2-amino-2-methyl-1-propanol, isobutanol-2-amine, 1-propanol, 2-amino-2-methyl, amp regular, 2-aminoisobutanol, aminomethylpropanol, 2-amino-2-methylpropanol, 2-methyl-2-aminopropanol, 2-aminodimethylethanol, hydroxy-tert-butylamine
11807
2-amino-2-methylpropan-1-ol
CC(C)(N)CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 31.0°C to 32.0°C
Color Colorless
Density 0.9300g/mL
Boiling Point 165.0°C
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3)2C(NH2)CH2OH
Refractive Index 1.4470 to 1.4490
Quantity 500 mL
Beilstein 04, III, 783
Fieser 01,37; 03,14; 06,20
Merck Index 15, 446
Specific Gravity 0.93
Solubility Information Solubility in water: miscible. Other solubilities: soluble in alcohols
Viscosity 102 mPa.s (30°C)
Formula Weight 89.14
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material 2-Amino-2-methyl-1-propanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'