missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2-methyl-1,3-propanediol, 99%, Thermo Scientific Chemicals

Catalog Number p-56671
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
This item is not returnable. View return policy
115-69-5
C4H11NO2
105.14
MFCD00004678
UXFQFBNBSPQBJW-UHFFFAOYSA-N
2-amino-2-methyl-1,3-propanediol, ampd, aminoglycol, gentimon, isobutandiol-2-amine, aminomethyl propanediol, ammediol, 1,1-di hydroxymethyl ethylamine, 1,3-propanediol, 2-amino-2-methyl, 2-amino-2-methyl-1,3-propandiol
1531
CHEBI:991
2-amino-2-methylpropane-1,3-diol
CC(CO)(CO)N
This item is not returnable. View return policy
115-69-5
C4H11NO2
105.14
MFCD00004678
UXFQFBNBSPQBJW-UHFFFAOYSA-N
2-amino-2-methyl-1,3-propanediol, ampd, aminoglycol, gentimon, isobutandiol-2-amine, aminomethyl propanediol, ammediol, 1,1-di hydroxymethyl ethylamine, 1,3-propanediol, 2-amino-2-methyl, 2-amino-2-methyl-1,3-propandiol
1531
CHEBI:991
2-amino-2-methylpropane-1,3-diol
CC(CO)(CO)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 108.0°C to 110.0°C
Color White
Boiling Point 151.0°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula HOCH2C(CH3)(NH2)CH2OH
Quantity 100 g
Beilstein 04, 303
Fieser 02,24
Merck Index 15, 445
Solubility Information Solubility in water: 2500g/L (20°C). Other solubilities: soluble in alcohol
Formula Weight 105.14
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-2-methyl-1, 3-propanediol, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'