missing translation for 'onlineSavingsMsg'
Learn More

(±)-2-Amino-1-phenylethanol, 95%, Thermo Scientific Chemicals

Catalog Number p-2689874
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
7568-93-6
C8H11NO
137.18
MFCD00008137
ULSIYEODSMZIPX-UHFFFAOYNA-N
phenylethanolamine, 2-hydroxy-2-phenylethylamine, phenethanolamine, beta-hydroxyphenethylamine, 2-phenyl-2-hydroxyethylamine, bisnorephedrine, beta-phenethanolamine, alpha-aminomethyl benzyl alcohol, 2-amino-1-phenylethan-1-ol, 2-amino-1-phenyl-1-ethanol
1000
CHEBI:16343
2-amino-1-phenylethanol
NCC(O)C1=CC=CC=C1
This item is not returnable. View return policy
7568-93-6
C8H11NO
137.18
MFCD00008137
ULSIYEODSMZIPX-UHFFFAOYNA-N
phenylethanolamine, 2-hydroxy-2-phenylethylamine, phenethanolamine, beta-hydroxyphenethylamine, 2-phenyl-2-hydroxyethylamine, bisnorephedrine, beta-phenethanolamine, alpha-aminomethyl benzyl alcohol, 2-amino-1-phenylethan-1-ol, 2-amino-1-phenyl-1-ethanol
1000
CHEBI:16343
2-amino-1-phenylethanol
NCC(O)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 56.0°C to 60.0°C
Color Yellow
Boiling Point 160.0°C (17.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula H2NCH2CH(C6H5)OH
Refractive Index 1.5580 to 1.5600
Quantity 10 g
Merck Index 15, 7399
Formula Weight 137.18
Percent Purity 98%
Physical Form Viscous Liquid
Chemical Name or Material (±)-2-Amino-1-phenylethanol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • (#1)-2-Amino-1-phenylethanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'