missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-1-cyclopentene-1-carbonitrile, 98%, Thermo Scientific Chemicals

Catalog Number p-7056966
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
2941-23-3
C6H8N2
108.144
MFCD00517551
NSMYBPIHVACKQG-UHFFFAOYSA-N
2-amino-1-cyclopentene-1-carbonitrile, 2-aminocyclopent-1-enecarbonitrile, 2-aminocyclopent-1-ene-1-carbonitrile, 1-amino-2-cyano-1-cyclopentene, 1-cyclopentene-1-carbonitrile, 2-amino, unii-u54887v9pj, 2-amino-cyclopent-1-enecarbonitrile, 1-amino-2-cyanocyclopent-1-ene, 1-amino-2-cyano-1,2-cyclopentene, 1-cyclopentene-1-carbonitrile, 2-amino-6ci,7ci,8ci,9ci
287275
2-aminocyclopentene-1-carbonitrile
C1CC(=C(C1)N)C#N
This item is not returnable. View return policy
2941-23-3
C6H8N2
108.144
MFCD00517551
NSMYBPIHVACKQG-UHFFFAOYSA-N
2-amino-1-cyclopentene-1-carbonitrile, 2-aminocyclopent-1-enecarbonitrile, 2-aminocyclopent-1-ene-1-carbonitrile, 1-amino-2-cyano-1-cyclopentene, 1-cyclopentene-1-carbonitrile, 2-amino, unii-u54887v9pj, 2-amino-cyclopent-1-enecarbonitrile, 1-amino-2-cyanocyclopent-1-ene, 1-amino-2-cyano-1,2-cyclopentene, 1-cyclopentene-1-carbonitrile, 2-amino-6ci,7ci,8ci,9ci
287275
2-aminocyclopentene-1-carbonitrile
C1CC(=C(C1)N)C#N

2-Amino-1-cyclopentene-1-carbonitrile s an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-1-cyclopentene-1-carbonitrile s an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Incompatible with heat and oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 147°C to 148°C
Color White
Quantity 25 g
UN Number UN3439
Beilstein 742061
Solubility Information Slightly soluble in water.
Formula Weight 108.14
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Amino-1-cyclopentene-1-carbonitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'