missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-1-cyclopentene-1-carbonitrile, 98%, Thermo Scientific Chemicals
242.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Description
2-Amino-1-cyclopentene-1-carbonitrile s an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Amino-1-cyclopentene-1-carbonitrile s an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Incompatible with heat and oxidizing agents.
2-Amino-1-cyclopentene-1-carbonitrile s an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Incompatible with heat and oxidizing agents.
Specifications
Melting Point | 147°C to 148°C |
Color | White |
Quantity | 100 g |
UN Number | UN3439 |
Beilstein | 742061 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 108.14 |
Percent Purity | 98% |
Physical Form | Solid |
Chemical Name or Material | 2-Amino-1-cyclopentene-1-carbonitrile |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'