missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-1,3-propanediol, 98%, Thermo Scientific Chemicals

Catalog Number 10508322
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
534-03-2
C3H9NO2
91.11
MFCD00051487
KJJPLEZQSCZCKE-UHFFFAOYSA-N
2-amino-1,3-propanediol, serinol, 1,3-propanediol, 2-amino, 2-amino-1,3-propandiol, 2-aminopropan-1,3-diol, unii-ic94l30j8m, 2-amino-1,3-dihydroxypropane, 2-amino-1,3-propanediol serinol, dl-serinol, sel
68294
2-aminopropane-1,3-diol
NC(CO)CO
This item is not returnable. View return policy
534-03-2
C3H9NO2
91.11
MFCD00051487
KJJPLEZQSCZCKE-UHFFFAOYSA-N
2-amino-1,3-propanediol, serinol, 1,3-propanediol, 2-amino, 2-amino-1,3-propandiol, 2-aminopropan-1,3-diol, unii-ic94l30j8m, 2-amino-1,3-dihydroxypropane, 2-amino-1,3-propanediol serinol, dl-serinol, sel
68294
2-aminopropane-1,3-diol
NC(CO)CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 52.0°C to 56.0°C
Color White
Boiling Point 115.0°C to 116.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula NH2CH(CH2OH)2
Quantity 1 g
Formula Weight 91.11
Percent Purity 98%
Physical Form Adhering Crystalline Powder
Chemical Name or Material 2-Amino-1, 3-propanediol
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • 2-Amino-1,3-propanediol
Signalwort
  • Danger
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Gefahrenhinweis
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'